Learn More
Abcam Ponasterone A, Ecdysone Analog, >95% Purity, 1 mg
Description
- Molecular Weight: 464.6 Da
- Purity: >95%
- Description: Ecdysone analog, a potent ecdysteroid activity inducer (EC50 = 70 nM). Increases β-galactosidase expression levels similar to Muristerone A and induces ecdysone-inducible mammalian expression systems in vivo.
- Chemical Formula: 100% Ponasterone A
- Storage Temperature: -20°C
- Shipping Temperature: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Buffer | 100% Ponasterone A |
| Content And Storage | Store at -20°C. |
| Shipping Condition | Ambient |
| Target Molecule | Ecdysteroid Receptor |
| Quantity | 1 mg |
| For Use With (Application) | Ecdysone-inducible mammalian expression system |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.