Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Ponasterone A, Ecdysone Analog, >95% Purity, 25 mg

Catalog No. 502705876
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5876 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5876 Supplier Abcam Supplier No. AB14433025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent ecdysteroid activity inducer with >95% purity.

  • Molecular Weight: 464.6 Da
  • Purity: >95%
  • Description: Ecdysone analog and potent ecdysteroid activity inducer (EC50 = 70 nM).
  • Function: Increases β-galactosidase expression levels similar to Muristerone A and induces ecdysone-inducible mammalian expression systems in vivo.
  • Storage: Store at -20°C.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Ecdysone Receptor
Shipping Condition Ambient
Buffer 100% Ponasterone A
Content And Storage Store at -20°C.
Formulation 100% Ponasterone A
Quantity 25 mg
For Use With (Application) Induces ecdysone-inducible mammalian expression system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.