Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Ponasterone A, Ecdysone Analog, >95% Purity, 5 mg

Catalog No. 502705875
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5875 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5875 Supplier Abcam Supplier No. AB1443305MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent ecdysteroid activity inducer used to trigger ecdysone-inducible mammalian expression systems.

  • Purity: >95%
  • Molecular Weight: 464.6 Da
  • CAS Number: 13408-56-5
  • Activity: Potent ecdysteroid activity inducer (EC50 = 70 nM)
  • Key Application: Induces ecdysone-inducible mammalian expression system in vivo; increases β-galactosidase expression levels

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Ecdysteroid Receptor
Substrate Properties Chemical Substrate
Buffer 100% Ponasterone A
Form Powder
Shipping Condition Ambient
Content And Storage Store at -20°C.
Quantity 5 mg
For Use With (Application) Induces ecdysone-inducible mammalian expression system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.