Learn More
Medchemexpress LLC HY-12489 500mg Medchemexpress, Ponceau S CAS:6226-79-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12489500MG
Encompass_Preferred
Medchemexpress, HY-12489 500mg Ponceau S CAS:6226-79-5 Ponceau S (Acid Red 112) is the most commonly used stain for Western blotting. Ponceau S is applied as an acidic aqueous solution. Ponceau S is compatible with antibody-antigen binding, and stains the proteins on the membrane red. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1058
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.