Learn More
Applied Biosystems™ POP-7™ Polymer for 3730/3730xl DNA Analyzers

Description
Includes
Each bottle contains enough polymer for 960 samples
- A separation matrix for the 3730 and 3730xl DNA Analyzers
- Customized for DNA sequencing and fragment analysis.
- Pre-formulated to save time and reproducibility by ensuring consistent formulation
- Designed to allow capillaries to be used multiple times for sequencing and fragment analysis applications
- Not suitable for use with ABI PRISM™, 310 and 3100 series genetic analyzers as it causes syringe failure and premature array degradation
- Sufficient for 8,000 to 12,000 samples
Use With
- 3730/3730xl Genetic Analyzer Capillary Array, 50 cm
- 3730 Running Buffer, 10X
- BigDye™ Terminator v1.1 Cycle Sequencing Kit
- BigDye™ Terminator v3.1 Cycle Sequencing Kit
- Hi-Di™ Formamide
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Accuracy | 98.5% up to 900 bases (for 50 cm capillary), 98.5% up to 700 bases (for 36 cm capillary) |
| Content And Storage | One bottle. Store at 2°C to 8°C. |
| For Use With (Equipment) | 3730xl DNA Analyzer, 3730 DNA Analyzer |
| For Use With (Application) | Standard Sequencing, Fragment Analysis (Denaturing) |
| On Instrument Life Usage | Stability on instrument 7 days, up to 12,000 samples/28 mL |
| Product Line | POP-7 |
| Product Type | Polymer |
| Quantity | 5 x 28 mL |
| Test Time | 1 hr for 36 cm capillary (std. seq.), 2 hrs for 50 cm capillary (long read seq.) |
Frequently Asked Questions (FAQs)
You can run 8,000-12,000 samples. These numbers may be lower if there is increased polymer consumption due to additional wizards being performed, such as the Bubble Remove and Water Wash wizards.
POP-7 polymer can maintain its performance for seven days on the Applied Biosystems 3730 Series DNA Analyzer at standard lab conditions. After seven days, perform all required maintenance (including the Water Wash Wizard) and replace with fresh polymer. Do not "top off" the polymer bottle (add fresh polymer to old polymer), which can adversely affect data quality and consumable lifespan.
Applied Biosystems supports the use of POP-7 polymer on the Applied Biosystems 3730 Series DNA Analyzer. If the instrument is running Applied Biosystems 3730 Data Collection Software v.3.1 or later, then you can also use POP-6 polymer. We do not recommend mixing and matching of various instrument polymers as each is manufactured in different processes, tested as an entire system, and guaranteed to perform to a minimum specification. Substitution of reagents may compromise the integrity of the system and cannot be supported by AB service or applications support specialists. Using unsupported polymers may adversely affect data quality, consumable lifespan, and may void existing warranties.
When placing the array on the instrument, the last step in the process is to loosen the array knob and the instrument forces any air bubbles around the array tip through the threading and the middle of the array knob. Some polymer gets pushed through as well and when it dries up, it appears as a white, flaky residue. Use a damp, lint-free cloth to remove it. If more residue appears after the cleanup, it is possible that the array knob is not tightened enough or the array ferrule may require replacement.
It is not unusual to see a large bubble at the tip of the piston while performing the water wash wizard on the system. When the polymer is put back on to the system, the difference in viscosity is usually enough to dislodge the bubble and push it out. In extreme cases, it may be necessary to repeat the wash and go back and forth between the water and polymer to force it out.
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.