Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Porcelain Mortar and Pestle Set
SDPSCI_ED

Porcelain Mortar and Pestle Set is neatly glazed for a consistent finish.

Supplier:  United Scientific Supplies JMD275

 View more versions of this product

Catalog No. S82842


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

  • Deep form mortars with lips are glazed on the outside (except for the bottom) and unglazed inside.
  • Oversized pestles are glazed to grinding surface.
  • Includes both mortars and pestles
  • Available in 7 sizes ranging from 50 mL to 1900 mL.
Specifications

Specifications

Glazed Outer, Unglazed Inner
275 mL
Mortar and Pestle
Porcelain
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.