Learn More
Eisco™ Porcelain Spot Plate
SCI_ED
Catalog No. S27076 Shop All Eisco Products
Change view
Click to view available options
No. of Wells:
6
12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells
S27076 6
S26588 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27076 Supplier Eisco™ Supplier No. CH0542A
Add to Cart
Add to Cart
White porcelain spot plates are deal for observing reactions with color changes.
  • Spot plate made of porcelain
  • Available with six or twelve wells
  • White for good visibility of color change reactions
  • Perfect for any laboratory
Specifications
Color White
Product Type Spot Plate
No. of Wells 6
Material Porcelain