Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Educational Products Porcelain Streak Plates
SCI_ED

Catalog No. S45052B Shop All American Educational Products LLC Products
Change view
Click to view available options
Quantity:
10/Pk.
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color
S45052B 10/Pk. Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S45052B Supplier American Educational Products Supplier No. 9215

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hard, unglazed porcelain displays true colors.

  • L x W: 1 x 2 in. (2.54 x 5.1cm)
  • Grades 4 to 12

Specifications

Quantity 10/Pk.
Grade 6 to 12
For Use With (Application) For rock, mineral, and fossil identification
Product Type Streak Plate
Includes Streak plate, Glass plate, hand lens, dropper bottle, magnet, nail, penny, harness scale and Ziplock pouch
Color Black
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.