Learn More
Alpco Ltd Porcine Albumin ELISA
SDP
Supplier:  Alpco Ltd 41ALBPOE01
Encompass
The Procine Albumin ELISA is a highly sensitive two-site enzyme linked immunoassay for measuring Albumin in porcine serum, tissue extracts and other biological fluids. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2885139