Learn More
Description
Specifications
Specifications
| Concentration | 1.0 mg/ml |
| For Use With (Application) | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Formulation | 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 |
| Quantity | 1 mg |
| Storage Requirements | Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles. |
| Regulatory Status | RUO |
| Cross Reactivity | Porcine IgG, Fab |
| Species | Porcine |
| Product Line | Novus |
| Purity or Quality Grade | Immunoelectrophoresis (IEP) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
