Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Porcine IL-2, AOF Recombinant Protein

Catalog No. PIRP87805
Encompass
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP87805 1 mg
PIRP87803 100 μg
PIRP87807 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PIRP87805 Supplier Gibco™ Supplier No. RP87805
Add to Cart
Add to Cart
Request a Quote

Recombinant Protein

Animal Origin Free Reconstitute with sterile water at 0.1 mg/mL.

Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P26891
Concentration 0.1 mg/mL
For Use With (Application) Control
Formulation 10 mM sodium phosphate with no preservative; pH 7.5
Gene ID (Entrez) 396868
Molecular Weight (g/mol) 15.3 kDa
Name Porcine IL-2, AOF
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration ≤1 EU/ μg
Gene Alias aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF
Common Name IL-2
Gene Symbol IL2
Biological Activity ED50 ≤ 1.0 ng/mL; determined by the dose-dependent proliferation of CTLL-2 cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MAPTSSSTKN TKKQLEPLLL DLQLLLKEVK NYENADLSRM LTFKFYMPKQ ATELKHLQCL VEELKALEGV LNLGQSKNSD SANIKESMNN INVTVLELKG SETSFKCEYD DETVTAVEFL NKWITFCQSI YSTLT
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade >95% by SDS-Page
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.