Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Porcine IL-6 Quantikine ELISA Kit
SDP

Catalog No. P6000B Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P6000B 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. P6000B Supplier R&D Systems™ Supplier No. P6000B

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Porcine IL-6 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Porcine IL-6 Quantikine ELISA Kit quantifies porcine IL-6 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).

Specifications

Assay Range 18.80 to 1200 pg/mL
Assay Sensitivity 4.3 pg/mL
Conjugate HRP
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (EDTA, Heparin), Plasma (Heparin), Serum, Serum
Specificity Natural and Recombinant Porcine IL-6
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 3569
Gene Symbol IL6
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2 A
Target IL-6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.