Learn More
ATCC Porcine parvovirus NADL-2 (ATCC VR-742) - frozen viral reference, BSL-2
Supplier: ATCC VR742
Porcine parvovirus NADL-2 (ATCC VR-742) is a reference strain used in virology and veterinary research. The complete genome (4,783 nt) is published to ATCC's genome portal. The material is supplied frozen and must be handled under Biosafety Level 2 conditions.
- Strain: NADL-2.
- Genus: Protoparvovirus.
- Genome length: 4,783 nt.
- Isolation source: porcine leukocytes.
- Format: Frozen viral material.
- Storage: -80°C.
- Biosafety level: BSL-2.
- Intended use: Virology and genomic research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.