Learn More
Thermo Scientific™ POROS™ MabCapture™ A Select Resin
Description
POROS™ MabCapture™ A Select Resin is a new high-throughput, lower-priced Protein A chromatography resin, combining past improvements to MabCapture A with our own Protein A ligand, enabling the same high-throughput purification performance at a significant reduction in price over most Protein A chromatography resins. These advances provide a monoclonal antibody purification resin that demonstrates high dynamic binding performance at low linear flow rates and maintains superior binding capacity at high linear flow rates where other chromatography media products cannot operate.
- Maintain high dynamic binding capacity at linear flow rates over 700 cm/hr
- Design processes with greater flexibility (shorter bed heights, faster flow rates, or both) due to the improvements made to the antibody binding efficiency
- Obtain the highest dynamic binding capacity for antibodies vs. conventional agarose- or silica-based Protein A resins
- Purify for polyclonal IgG from human, rabbit, monkey, mouse, pig, dog, or cat serum
- Clean and sanitize with standard sodium hydroxide agents used for other chromatography products
- Recognize major increases to process productivity and reduce the downstream bottleneck
The Protein A molecule contains five high-affinity binding sites capable of binding to the Fc portion of antibodies. Compared to Protein G, Protein A provides the highest binding capacity for subclasses of IgG from rabbits, pigs, dogs, and cats. Protein A also has higher binding capacity for human IgG, except for IgG3, which it binds weakly. Protein A also binds weakly to mouse IgG1 and is not recommended for most applications with that antibody isotype. The Protein A molecule also has greater stability than Protein G. Protein A is heat-stable and retains its native conformation when exposed to denaturing reagents such as 4M urea, 4M thiocyanate, and 6M guanidine hydrochloride.
The development of humanized monoclonal antibodies has continued to fuel the growth of the biotherapeutic market. Addressing the therapeutic demand for monoclonal antibodies has led to major improvements to the cell-based expression systems that are used to manufacture these proteins. >
Consequently, these improvements have moved the processing bottleneck from upstream cell expression to downstream purification. Protein A chromatography, which is used as the primary capture chromatography step in most antibody process platforms, is generally regarded as the major process bottleneck due to the slow flow rates and long residence times required for the effective operation of conventional Protein A chromatography resin. POROS MabCapture Select chromatography resin offers the capacity, efficiency, and flexibility needed to make improvements to the downstream purification platform, resulting in major increases to process productivity and antibody manufacturing
Specifications
Specifications
| Type | A Select Resin |
| Standard Components | Protein A |
| For Use With (Application) | IgG Purification |
| Volume (Metric) | 5 mL |
| Content And Storage | Store in refrigerator (2–8°C). |
| Product Line | POROS |
| Format | Poros Bead |
| Stationary Phase | Protein A |
| Purification Target | IgG Antibodies, Ig Antibodies |
| Shipping Condition | Wet Ice |
Frequently Asked Questions (FAQs)
All of the pre-packed POROS columns, regardless of column size or resin, have the same 10-32 female fittings on the column. Columns are compatible with Waters and Agilent HPLC systems. You will need 1/16 inch male fittings for the HPLC to attach the column.
All of the pre-packed columns, regardless column size or resin, have the same 10-32 female fittings on the column. They are all compatible with the AKTA instrument platform, however due to the column volume of the 2.1 mmD x 30 mmL columns (0.1 mL), these will not perform well in AKTA or Avant purification systems due to the relative size of the hold- up volume in these systems. The smallest column (PEEK or SS) that could feasibly be used in these systems is the 4.6mmD x 50mmL (0.8 mL) format.
The physical stability of POROS resins is high. Customers have agitated, with overhead mixing and or bottom gassing for long periods of time (5-10 hours) and have performed hundreds of diaphragm pump passes during stall packs. Keep in mind that agitation with any device that creates grinding and sheer, such as with a magnetic stir bar, is not recommended for any type of resin.
POROS resins rehydrate quite well. Here is the procedure that we recommend:
1. Flow 5% ethanol over the column at ?350 cm/hour for 3 CVs, followed by water or 0.1 M sodium chloride at 300 cm/hour for 3 CVs.
2. If baseline noise is still present, flow 20% ethanol over the column at 300 cm/hour for 3 CVs, followed by water or 0.1 M sodium chloride at 300 cm/hour for 3 CVs.
3. If you have established column qualification tests and specifications, perform the tests after rewetting.
In general POROS resins have behaved better for large biomolecules such as viral particles, fusion proteins, and globular proteins, where shape (not size) matters due to its unique pore structure and large pore structure as compared to traditional resins. As the target molecule size increases, capacities obtained will decrease. The large pore structure of POROS resins, which allows for convective flow (and therefore enhanced diffusion) is especially well suited for the purification of large biomolecules. POROS resins offer the best blend of high capacity (associated with chromatography beads) and improved chromatography efficiency (typically associated with monoliths or membranes). The average pore size for POROS resins is 1,000-3,600 Angstrom (100-360 nm) depending on the base bead/chemistry being utilized. For example, the molecular weight of IgG is 150 kDa and the molecular radius is 55 Angstrom. The molecular weight of IgM is ~900 kDa (pentamer), and the molecular radius is 120 Angstrom. So both of these biomolecules can interact with the pore structure associated with POROS materials.
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.