Learn More
Thermo Scientific™ POROS™ 50 PI RoboColumn™ Pre-packed columns
Catalog No. A30721
Change view
Click to view available options
Column Capacity:
200 μL
600 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Column Capacity
A30721 200 μL
A30722 600 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30721 Supplier Thermo Scientific™ Supplier No. A30721
Only null left
Add to Cart
Add to Cart
Thermo Scientific™ RoboColumn™ chromatography products are small columns provided in 8-column strips.
RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS PI50 resin, a 50-micron weak anion-exchange resin suitable for low-pressure applications, including large-scale downstream bioprocessing.
POROS PI50 chromatography resin is based on a polyethyleneimine functional group, yielding a high capacity. It is designed for the separation and purification of biomolecules.
Features of the resin:
  • Higher productivity—through high throughput and high dynamic capacity associated with high performance chromatography
  • Chemical stability—allows aggressive cleaning and sanitization
  • Enhanced biomolecule access—provided via large pores (500-10,000 Å)
  • Polystyrenedivinylbenzene particles—yield a robust, easily packable matrix
  • Better separation methods in less time—the speed of high performance chromatography reduces weeks of experimentation to only a few hours, leaving more time to explore all the variables of your separation
RoboColumn™ is a registered trademark of ATOLL GmbH.
Specifications
Content And Storage Store at 2°C to 8°C or at room temperature.
Column Capacity 200 μL
Flow Rate 16 to 1000 cm/hr.
Quantity 200 μL