Learn More
Grainger PORT SURF ROUGH TSTER SJ-210
Supplier:  Grainger 5RCG9
Encompass_Preferred
Mitutoyo Portable Surface Roughness Tester with External Display: SJ-210, 5 um Probe Tip Radius. Portable surface roughness testers, also known as surface testers, are built with a probe that can extend from the body and digital display to reach large workpieces, hard-to-reach features, and take other measurements in complex positions. They can cycle through standard measurements such as Ra and Rz of a surface.
Catalog No. NC2184079
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.