Learn More
Honeywell Fendall Porta Stream™ III Eyewash Station
Description
- Sturdy, molded unit features simple, highly visible instructions for correct operation
- Unit includes a 180 oz. sperian saline concentrate that can be easily placed near most workplace hazards with single bracket mounting device
- Large fill hole allows refill of concentrate solution and simplifies cleaning
- Efficient full flow nozzle provides a wide area around the eye
Specifications
Specifications
| Capacity (English) | 16 gal. |
| Capacity (Metric) | 60.5 L |
| Certifications/Compliance | ANSI Z358.1 |
| Flow Rate | 0.40 gpm |
| Includes | 180 oz. Saline Solution, Nozzle Strap |
| Mounting | Single Bracket |
| Product Type | Porta Stream™ III Eyewash Station |
| Rinse Time | 15 min. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.