Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Portable Multi-Gas Calibration Gas Kit (Methane)

Catalog No. 19805704 Shop All 3M/Aerion Products
Change view
Click to view available options
For Use With (Equipment):
950 Series Multigas Detectors
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-805-704 950 Series Multigas Detectors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-805-704 Supplier 3M™ Supplier No. 933000023/DEL

missing translation for 'validMainframeMsg'

For use with 950 series Multi-gas detectors.

  • Capacity: 34L
  • Composition: 10% LEL ME/35 PPM CO/10 PPM HS/18% OX
  • Flow Rate: 0.2 LPM, 715 Regulator

Specifications

For Use With (Equipment) 950 Series Multigas Detectors
Type Portable Multi-Gas Calibration Gas Kit (Methane)
For Use With (Application) Calibration of 950 series Multi-gas detectors
Includes 10% LEL ME/35 PPM CO/10 PPM HS/18% OX
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.