Learn More
Cole-Parmer™ Oakton™ Portable Turbidity Meter Kit
Catalog No. 13000102 Shop All Oakton Products
Change view
Click to view available options
Light Source:
Infrared
White Light
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Light Source
13-000-102 Infrared
13-000-101 White Light
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-000-102 Supplier Cole-Parmer™ Supplier No. 3563515
Add to Cart
Add to Cart
Provides accurate results with simple calibration and testing
  • Auto-ranging from 0 to 1000 NTU (FNU)
  • Stores up to 200 sets of data
  • Suitable for field use
  • Meets IP67 water-resistant requirements
Specifications
Type Turbidity Meter
Light Source Infrared
Display Type Color TFT
Response Time Variable
Automatic Range Selection Yes
Test Reagent Formats Liquid
Key Functions Measuring, Calibration, Parameter Setup, Data Recall
IP Rating IP67
Precision ≤ ±2% of reading stray light
Resolution 0.01/0.1/1 NTU
Turbidity 0 to 1000 NTU
Battery Life Dependent on use
Battery Type AA batteries
EPA Approved Methods None
ISO Accepted Methods ISO 7027 (DIN EN 27027) method
Warranty 2 Years