Learn More
Apexbio Technology LLC Posaconazole, 171228-49-2, MFCD00941162, 5 mg
SDP
Supplier:  Apexbio Technology LLC A17185
Encompass_Preferred
MF: C37H42F2N8O4, MW: 700.78, Solubility: 35.039mg/mL in DMSO, Appearance: A solid. Posaconazole, also known as SCH 56592 or Noxafil, is a novel triazole antifungal compound that potently inhibits 14alpha-demethylation in a variety of fungi. Posaconzole, an itraconzole derivative with fluorine replacing chlorine in the phenyl ring and hydroxylation in the side chain, binds to the heme cofactor on the active site of 14alpha-demethylase, which converts lanosterol to 14alpha-dimethy lanosterol, resulting in the disruption of the integrity and function of the fungal cell membrane and inhibition of fungal growth.
Catalog No. 50-101-1940
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.