Learn More
MEDTOX™ Positive Control
Description
- MEDTOX 300 Positive Toxicology Control: Use for Opiate 300 tests (PROFILE-II ER and all VERDICT -II products)
- MEDTOX 2000 Positive Toxicology Control: Use for Opiate 2000 tests (All PROFILE-II except ER)
- Positive control urines have been spiked with authentic reference drug standards and/or appropriate metabolites
- Should be treated as any “unknown” specimen while following the specific protocol of the assay being used
- Controls prepared from human based urine available as a negative and Cutoff +50% to monitor the performance of MEDTOX drugs of abuse test devices
- Each bottle contains stabilized human based urine
Unopened:
The controls are stable for two years or until the expiration date, whichever comes first, when stored at -20°C protected from light.
The controls are stable for six months or until the expiration date, whichever comes first, when stored at 2 to 8°C
After Opening:
The controls are stable for six months or until the expiration date, whichever comes first, when stored at -20°C (Controls can be aliquoted and frozen)
The controls are stable for one month or until the expiration date, whichever comes first, when stored at 2 to 8°C
Specifications
Specifications
| Sample Type | Urine |
| Type | MEDTOX 300 Positive Toxicology Control |
| Intended Use | Positive control used for quality control with MEDTOX PROFILE-II ER and all VERDICT -II products |
| Packaging | Bottle |
| Quantity | 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.