Learn More
ACROBiosystems Post-Fusion glycoprotein 25ug
Supplier:  ACROBiosystems PONV82E325UG
Encompass_Preferred
Biotinylated Nipah virus Post-Fusion glycoprotein HisAvitag (PON-V82E3) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Asn 99 & Gly 117 - Ser 488 (Accession # Q9IH63-1).
Catalog No. 50-253-4295
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.