Learn More
Surgical Design™ Post Mortem Hammer Hook Blade
Catalog No. 21124628 Shop All Surgical Design Products
Change view
Click to view available options
Length (English):
9.5 in.
Catalog No. Length (English)
21-124-628 9.5 in.
1 options
Catalog No. 21-124-628 Supplier Surgical Design™ Supplier No. ATPM008
Add to Cart
Add to Cart
Post mortem hammer, 9.5 in., with integrated hook blade supports lifting, retracting, and controlled bone work during cranial and thoracic examinations.
  • Dual-function design combines a solid stainless-steel hammer head with a curved hook blade for efficient autopsy workflow
  • 9.5-in. length provides leverage and control for cranial, thoracic, and skeletal examinations
  • Hammer head is engineered for precise tapping and controlled bone separation, commonly used during skullcap removal
  • Curved hook blade assists in lifting soft tissue, securing edges of incisions, and retracting structures without slippage
  • Constructed from premium surgical-grade stainless steel for durability in high-intensity postmortem use
  • Single-piece forged design ensures strength, balance, and longevity
  • Satin-finish handle for a reliable, non-slip grip, even with gloved or wet hands
  • Fully autoclavable and corrosion resistant, suitable for routine hospital pathology and forensic labs
  • Ideal for brain removal protocols, neck dissections, thoracic cavity access, and general postmortem tissue handling
  • Assisting with cranial autopsy, including lifting the skull cap after oscillating saw cuts
  • Retracting soft tissue during thoracic or cervical dissection
  • Providing leverage for bone plate removal and repositioning
  • Useful in forensic, pathology, medical examiner, and teaching laboratory settings
Specifications
Product Type Hammer
Material Stainless Steel
Length (English) 9.5 in.
Sterility Non-sterile