Learn More
Med Vet International Renaplus Gel, 5oz
Supplier:  Med Vet International RXRENAPLUSGEL
Encompass_Preferred
RenaPlus Gel is a prescription potassium gluconate supplement that is used for Hypokalemia or low blood potassium levels in cats and dogs.
Catalog No. 50-282-7887
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.