Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium 3-Pyridyltrifluoroborate 98.0+%, TCI America™
SDP

Catalog No. P17171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17171G 1 g
P17175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17171G Supplier TCI America Supplier No. P17171G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 561328-69-6
Molecular Formula C5H4BF3KN
Molecular Weight (g/mol) 185.00
MDL Number MFCD09038545
InChI Key MNCKCIOWIHHCSC-UHFFFAOYSA-N
PubChem CID 23692458
IUPAC Name potassium trifluoro(pyridin-3-yl)boranuide
SMILES [K+].F[B-](F)(F)C1=CN=CC=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 185.00
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Potassium 3-Pyridyltrifluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.