Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Acetate ACS AR Crystal, Macron Fine Chemicals™

Catalog No. MK6700500 Shop All Avantor Products
Change view
Click to view available options
POLYSTORMOR; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-6700-500 POLYSTORMOR; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-6700-500 Supplier Avantor Supplier No. 670004

Specifications

CAS 127-08-2
Physical Form Crystalline
Quantity 500 g
Molecular Formula C2H3KO2
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES CC(=O)[O-].[K+]
IUPAC Name potassium;acetate
Molecular Weight (g/mol) 98.142
PubChem CID 517044
ChEBI CHEBI:32029
Formula Weight 98.14
Grade ACS
Packaging POLYSTORMOR
Chemical Name or Material Potassium Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.