Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Acetate, Crystalline Powder, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605434 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-434 12 kg Poly Pail
18-605-429 125 g Poly Bottle
18-605-430 500 g Poly Bottle
18-605-432 2.5 kg Poly Pail
18-605-431 6 x 500 g Poly Bottle
18-605-433 4 x 2.5 kg Poly Pail
18-605-435 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-605-434 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PO15512KG
Add to Cart
Add to Cart

PO155, 127-08-2, C2H3KO2

Spectrum™ Chemical Potassium Acetate, Crystalline Powder, USP is a potassium salt used to replenish electrolytes, for restoration of water-electrolyte balance, as well as, a urinary and systemic alkalizer. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.14
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
IUPAC Name potassium acetate
SMILES [K+].CC([O-])=O

Specifications

pH 7.5 to 8.5
CAS Max % 1
Quantity 12 kg
Percent Purity 99 to 100.5%
Grade USP
Loss on Drying 0.01
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.