Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Acid Phthalate, 0.0500 N (N/20), Ricca Chemical
SDP

Catalog No. 58701 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58701 4 L
5870-16 500 mL
5870-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 58701 Supplier Ricca Chemical Company Supplier No. 58701
Add to Cart
Add to Cart

For Precision Titrations Testing

Specifications

Chemical Name or Material Potassium Acid Phthalate
Color Colorless
CAS 877-24-7, 7732-18-5
CAS Min % 97.0, 0.99
CAS Max % 100.0, 1.03
Molecular Formula C8H5KO4
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
IUPAC Name potassium;2-carboxybenzoate
Molecular Weight (g/mol) 204.222
PubChem CID 23676735
Grade Laboratory
Packaging Natural Poly Bottle
pH ∼4
Quantity 4 L
Physical Form Liquid
Concentration or Composition Notes 0.0500Normal
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.