Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bicarbonate, ≥99.95% Trace Metals Basis, Solstice

Catalog No. 60046869 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-869 250 g Poly Bottle
60-046-870 50 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-869 Supplier Solstice Supplier No. 431583250G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

≥99.95% trace metals basis, 99.7-100.5% dry basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
IUPAC Name potassium;hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

Melting Point 100°C to 120°C
Color White
pH 8 to 8.6 (at 20°C)
Quantity 250 g
Linear Formula KHCO3
UN Number NONH for all modes of transport
Beilstein 4535309
Solubility Information In water 333g/L at 20°C
Formula Weight 100.12g/mol
Percent Purity ≥99.95% (trace metals basis)
Odor Odorless
Packaging Poly Bottle
Appearance Powder
Density 2.17g/cm3 at 20°C
Chemical Name or Material Potassium bicarbonate
Show More Show Less
RTECSNumber 206-059-0
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.