Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Biphthalate, BiotechGrade, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18605463 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
6 x 500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-463 500 g Poly Bottle
18-605-464 6 x 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-605-463 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1192500GM
Only null left
Add to Cart
Add to Cart

P1192, 877-24-7, C8H5O4K

Spectrum™ Chemical Potassium Biphthalate, BiotechGrade , also known as potassium hydrogen phthalate or KHP, is an acidic salt compound used as a primary standard for acid-base titrations and as a buffering agent. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.22
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
IUPAC Name potassium 2-carboxybenzoate
SMILES [K+].OC(=O)C1=CC=CC=C1C([O-])=O

Specifications

pH 3.5 to 5.0
CAS Max % 1
Quantity 500 g
Percent Purity 99.5%
Grade Biotech
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.