Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bisulfate, Puriss. p.a., ≥99%, Solstice

Catalog No. 6001195 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-011-95 500 g
60-011-94 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-011-95 Supplier Solstice Supplier No. 31260500G

Puriss. p.a.,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-93-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
MDL Number MFCD00011404
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
Synonym potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
PubChem CID 516920
IUPAC Name potassium;hydrogen sulfate
SMILES [K+].OS([O-])(=O)=O

Specifications

Quantity 500 g
Linear Formula KHSO4
UN Number 2509
Formula Weight 136.17g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Density 2.32g/mL at 25°C
Chemical Name or Material Potassium Bisulfate
Hazard Statement P261-P280-P305 + P351 + P338-P310
Precautionary Statement H314-H335
EINECSNumber 231-594-1
RTECSNumber TS7200000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.