Learn More
Potassium bromate, 99%
Description
Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.
Solubility
Slightly soluble in alcohol but insoluble in acetone and ethanol.
Notes
Strong oxidizer. Incompatible with combustible materials, organics, reducing agents, aluminum and finely powdered metals
Specifications
Specifications
| Melting Point | ∼370°C (decomposition) |
| Quantity | 2500 g |
| UN Number | UN1484 |
| Merck Index | 14,7617 |
| Solubility Information | Slightly soluble in alcohol but insoluble in acetone and ethanol. |
| Formula Weight | 167 |
| Percent Purity | 99% |
| Odor | Odorless |
| Density | 3.27 |
| Chemical Name or Material | Potassium bromate |
Safety and Handling
| Hazard Category | H271-H301-H335-H336-H351-H373 |
| Hazard Statement | GHS H Statement H271-H301 May cause fire or explosion; strong oxidizer. Toxic if swallowed. |
| Precautionary Statement | P201-P202-P210-P220-P221-P260-P264b-P270-P271-P280-P283-P301+P310-P304+P340-P306+P360-P308+P313-P330-P370+P378q-P371+P380+P375-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: POTASSIUM BROMATE |
| EINECSNumber | 231-829-8 |
| RTECSNumber | EF8725000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.