Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bromate RV, EP Primary Standard, Ricca Chemical
SDP

Catalog No. 58794500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58794-500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 58794-500 Supplier Ricca Chemical Company Supplier No. 5879.4500
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7758-01-2
Molecular Formula BrKO3
Molecular Weight (g/mol) 167.00
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
Synonym potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
PubChem CID 23673461
ChEBI CHEBI:38211
IUPAC Name potassium;bromate
SMILES [O-]Br(=O)=O.[K+]

Specifications

Color White
CAS Min % 65.0, 32.68
CAS Max % 68.0, 34.01
Physical Form Granular/Lump
Quantity 500 g
Percent Purity 100%
Grade EP
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Potassium Bromate RV, Primary Standard
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.