Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bromide, 10% (w/v) Aqueous, Ricca Chemical
SDP

Catalog No. p-2916814 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
58795-32 1 L Natural Poly Bottle
58795-1 4 L Natural Poly Bottle
5879525 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 58795-32 Supplier Ricca Chemical Company Supplier No. 5879.532
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Potassium Bromide
CAS 7758-02-3, 7732-18-5
Molecular Formula BrK
MDL Number MFCD00011358
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES [K+].[Br-]
IUPAC Name potassium bromide
Molecular Weight (g/mol) 119.00
PubChem CID 253877
ChEBI CHEBI:32030
Percent Purity 10%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
pH Neutral
Quantity 1 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.