Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bromide, Free-flowing, ACS reagent, anhydrous, ≥99%, Solstice

Catalog No. 60047504 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-504 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-504 Supplier Solstice Supplier No. 746444500G

Free-flowing, ACS reagent, anhydrous,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
IUPAC Name potassium;bromide
SMILES [K+].[Br-]

Specifications

Boiling Point 1,383°C
Melting Point 734°C (literature)
CAS Max % 100.0000%
Physical Form Anhydrous
Quantity 500 g
Linear Formula KBr
Formula Weight 119
Percent Purity >99%
Grade ACS Reagent
Packaging Poly Bottle
Density 2.73 g/cm3
Chemical Name or Material Potassium bromide, Free-flowing, ACS reagent, anhydrous, ≥99%
Show More Show Less
ShelfLife 720 days from date of manufacture
EINECSNumber 231-830-3
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.