Learn More
Potassium bromide, FT-IR Grade, ≥99% trace metals basis, Solstice
Shop All Honeywell Research Chemicals ProductsSpecifications
Specifications
| Quantity | 100 g |
| Linear Formula | KBr |
| UN Number | NONH for all modes of transport |
| IUPAC Name | potassium;bromide |
| Formula Weight | 119g/mol |
| Percent Purity | ≥99% (trace metals basis) |
| Packaging | Poly bottle |
| Vapor Pressure | 1mmHg (795°C), 0.01mmHg (20°C) |
| Chemical Name or Material | Potassium Bromide |
Safety and Handling
| Hazard Statement | P280-P305 + P351 + P338-P337 + P313 |
| Precautionary Statement | H319 |
| EINECSNumber | 231-830-3 |
| RTECSNumber | TS7650000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.