Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chlorate, Purified, Reagents

Catalog No. 01337117
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-117 500 g
01-337-118 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-117 Supplier Reagents Holdings Llc Supplier No. C2280000500A5
Add to Cart
Add to Cart

Potassium Chlorate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 3811-04-9
Molecular Formula ClKO3
Molecular Weight (g/mol) 122.55
InChI Key VKJKEPKFPUWCAS-UHFFFAOYSA-M
IUPAC Name potassium chlorate
SMILES [K+].[O-][Cl](=O)=O

Specifications

Boiling Point 400°C
Melting Point 368°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Poly Bottle
Specific Gravity 2.32
Chemical Name or Material Potassium Chlorate, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.