Learn More
Potassium chloride, 0.159M, Conductivity Standard
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
| Density | 1.01181g/cm3 at 20°C |
| Assay Percent Range | 0.159M |
| Physical Form | Liquid |
| Color | Colorless |
| IUPAC Name | potassium chloride |
| Quantity | 1 L |
| Formula Weight | 74.55 |
| Concentration or Composition (by Analyte or Components) | Potassium chloride: 1.2%; Water: 98.8% |
| Chemical Name or Material | Potassium chloride, 20, 000μmho at 25°C |
Safety and Handling
| EINECSNumber | 231-211-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.