Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, 2.00 M, Ricca Chemical
SDP

Catalog No. 589921 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
589921 4 L Natural Poly Bottle
5899216 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 589921 Supplier Ricca Chemical Company Supplier No. 5899.21
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7447-40-7

Specifications

Color Colorless
Physical Form Liquid
Quantity 4 L
Molecular Formula C2H3NaO2
MDL Number MFCD00012459
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
SMILES [Na+].CC([O-])=O
IUPAC Name sodium acetate
Molecular Weight (g/mol) 82.03
PubChem CID 517045
ChEBI CHEBI:32954
Grade Laboratory
Concentration or Composition (by Analyte or Components) 2.00 Molar
Packaging Natural Poly Bottle
Chemical Name or Material Potassium Chloride
Show More Show Less
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.