Learn More
Potassium Chloride Conductivity Standard, 1500 μS/cm at 25°C, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Packaging | Cubitainer |
| Odor | Odorless |
| Assay Percent Range | 1, 500 ± 1 μS/cm at 25°C |
| Physical Form | Liquid |
| Color | Colorless |
| pH | Approximately 7 |
| Boiling Point | 100°C |
| Solution Type | Conductivity Standard |
| Type | Conductivity Standard |
| Solvent or Matrix | Water |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.