Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride Conductivity Standard, 1000 μS/cm at 25°C, Ricca Chemical
GSA_VASDP

Catalog No. 5888015 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cube
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
58-880-15 20 L Cube
588801-16 500 mL Natural Poly Bottle
588801-32 1 L Natural Poly Bottle
5888011 4 L Natural Poly Bottle
R588801010F 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 58-880-15 Supplier Ricca Chemical Company Supplier No. R588801020F
Add to Cart
Add to Cart

Tested in ISO 17025 accredited laboratories, providing the utmost quality in calibration.

  • Tested in ISO 17025 Accredited Laboratories.
  • Manufactured in cGMP compliant, FDA registered facilities.
  • Certified Traceable to NIST Standard Reference Material when available.
  • Many C of As and labels contain TDS and conductance information.
  • Tightest Specifications in the industry, lowest lot-to-lot variability.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Packaging Cube
Certificate of Analysis Yes
Traceability to NIST SRM# 999
CAS Min % 99.95%, 0.05%
CAS 7732-18-5, 7447-40-7
CAS Max % 0.05%, 99.95%
Conductivity 999 to 1001 μS/cm
Technique Calibration of Conductivity Meter
Certifications/Compliance ISO 17025
Color Colorless
Boiling Point 100°C
For Use With (Equipment) Conductivity Meters and Electrodes
Solubility Miscible with water
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 20 L
Show More Show Less
ShelfLife 24 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.