Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride Conductivity Standard, 20,000 μS/cm (20 mS/cm) at 25°C, Ricca Chemical
SDP

Catalog No. R588820010F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R588820010F 10 L Cubitainer
R58882004A 4 L Natural Poly Bottle
R588820020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R588820010F Supplier Ricca Chemical Company Supplier No. 5888.202.5
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Cubitainer
Odor Odorless
Assay Percent Range 20.0 ± 0.1 mS/cm at 25°C
Physical Form Liquid
Color Colorless
pH Approximately 7
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Solvent or Matrix Water
Quantity 10 L
Chemical Name or Material Potassium Chloride Conductivity Standard, 20 mS/cm (20, 000μmho/cm) at 25°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.