Learn More
Potassium Chloride Conductivity Standard, 25,000 μS/cm (25 mS/cm) at 25°C, Ricca Chemical

Shop All Ricca Chemical Company Products


Description
- Tested in ISO 17025 Accredited Laboratories.
- Manufactured in cGMP compliant, FDA registered facilities.
- Certified Traceable to NIST Standard Reference Material when available.
- Many C of As and labels contain TDS and conductance information.
- Tightest Specifications in the industry, lowest lot-to-lot variability.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Packaging | Poly |
| Certificate of Analysis | Yes |
| Traceability to NIST | SRM# 999 |
| CAS Min % | 98.52%, 1.48% |
| CAS | 7732-18-5, 7447-40-7 |
| CAS Max % | 1.48%, 98.52% |
| Conductivity | 24900 to 25100 μS/cm |
| Technique | Calibration of Conductivity Meter |
| Certifications/Compliance | ISO 17025 |
| Color | Colorless |
| Show More |
Safety and Handling
| ShelfLife | 24 Months |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.