Learn More
Potassium Chloride Conductivity Standard, 5000 μS/cm at 25°C, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Packaging | Cubitainer |
| Odor | Odorless |
| Assay Percent Range | 5.00 ± 0.01 mS/cm at 25°C |
| Physical Form | Liquid |
| Color | Colorless |
| pH | Approximately 7 |
| Boiling Point | 100°C |
| Solubility | Infinitely soluble in water |
| Solution Type | Conductivity Standard |
| Type | Conductivity Standard |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.