Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, Crystal, ACS, 99.0-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18612549 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
113 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-549 12 kg Poly Pail
18-612-546 125 g Poly Bottle
18-612-547 500 g Poly Bottle
18-612-548 2.5 kg Poly Bottle
18-612-550 50 kg Fiber Drum
18-612-551 113 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-612-549 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P125012KG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

pH 5.4 to 8.6
CAS Max % 1
Quantity 12 kg
Percent Purity 99.0 to 100.5%
Grade ACS
Packaging Poly Pail

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.