Learn More
Specifications
Specifications
| Boiling Point | 1420°C |
| Melting Point | 770°C |
| Color | White |
| pH | 6 |
| Physical Form | Solid |
| Quantity | 250 lb. |
| Formula Weight | 74.55 |
| Percent Purity | 99.0 to 100.5% (USP) |
| Grade | BP/EP/FCC/USP |
| Identification | Pass Test (BP), Pass Test (EP), Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC/EP/BP) |
| Show More |
Safety and Handling
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen. NFPA Health:1 Flammability:0 Instability:1 |
| Recommended Storage | Keep containers tightly closed in a dry, cool and well-ventilated place. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.