Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Potassium Chloride, Molecular Biology Grade, Calbiochem™,

Catalog No. 5295521KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-955-21KG 1 kg
52-955-2250GM 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52-955-21KG Supplier MilliporeSigma™ Supplier No. 5295521KG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Quantity 1 kg
Assay Percent Range ≥99%
Solubility Information Easily soluble in cold water
Formula Weight 74.6
Chemical Name or Material Potassium Chloride
Grade by argentometry
RTECSNumber TS8050000
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.