Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, Saturated, Approximately 4 M, Ricca Chemical
SDP

Catalog No. 59001 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5900-1 4 L Natural Poly Bottle
5900-4 120 mL Natural Poly Bottle
5900-16 500 mL Natural Poly Bottle
5900-32 1 L Natural Poly Bottle
5900-25 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 5900-1 Supplier Ricca Chemical Company Supplier No. 59001
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Color Colorless
CAS Min % 75.0, 22.63
CAS Max % 78.0, 23.55
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Saturated Aqueous Solution
pH ∼7
Quantity 4 L
Physical Form Liquid With Possible Precipitate
Concentration or Composition Notes approximately 4Molar
Chemical Name or Material Potassium Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.