Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, Saturated at 20°C, Reagents

Catalog No. 01334352
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-352 500 mL
01-334-354 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-352 Supplier Reagents Holdings Llc Supplier No. CP113900500A
Add to Cart
Add to Cart

Potassium Chloride, Saturated at 20°C, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

Molecular Formula KCl
Molecular Weight (g/mol) 74.55
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Muriate of Potash
SMILES O

Specifications

Boiling Point 104°C
CAS 7732-18-5, 7447-40-7
Color Colorless
pH 5.4 to 8.6
Physical Form Liquid
Quantity 500 mL
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.1481
Chemical Name or Material Potassium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.