Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chromate (Granular/Certified ACS), Fisher Chemical™

Catalog No. P220100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P220-100 100 g Poly Bottle
P220-500 500 g Poly Bottle
P220-3 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P220-100

Chemical Identifiers

CAS 7789-00-6
Molecular Formula CrK2O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00011368
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
Synonym potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
PubChem CID 24597
ChEBI CHEBI:75249
IUPAC Name dipotassium dioxochromiumbis(olate)
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O

Specifications

Melting Point 975°C
Color Yellow
pH 8.6 to 9.8
Physical Form Solid
Quantity 100 g
Assay Percent Range 99.5 to 100.5 %
UN Number 3087
Formula Weight 194.19
Percent Purity 99.5 to 100.5%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Potassium Chromate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. May cause cancer by inhalation. May cause heritable genetic damage. Irritating to eyes, respiratory system and skin. May cause an allergic skin reaction. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:2
DOTInformation DOT Class 5.1, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.